Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: PAPAEPQPGG SQCVEHDCFA LYPGPATFLN ASQICDGLRG HLMTVRSSVA ADVISLLLNG DGGVGRRRLW IGLQLPPGCG DPKRLGPLRG FQWVTGDNNT SYSRWARLDL NGAPLCGPLC VAVSAAEATV PSEPIWEEQQ CEVKADGFLC EFHFPATCRP LAVEPGAAAA AVSITYGTPF AARGADFQAL PVGSSAAVAP LGLQLMCTAP PGAVQGHWAR EAPGAWDCSV ENGGCEHACN AIPGAPRCQC PAGAALQADG RSCTASATQS CNDLCEHFCV PNPDQPGSYS CMCETGYRLA ADQHRCEDVD DCILEPSPCP QRCVNTQGGF ECH
Thrombomodulin (TM) is also known as Fetomodulin (FM), endothelial anticoagulant protein, or glycoprotein P112. Thrombomodulin contains six repeated domains homologous with epidermal growth factor (EGF) and an amino terminal domain homologous to lectin-like protein. Through its accelerated activation of protein C, synthesis of TM is one of several mechanisms important in maintaining thrombo-resistance and thus reducing clot formation on the surface of the endothelial cells. Immunohistochemical staining of formalin-fixed tissue for TM has been used for the study of a variety of vascular tumors, meningiomas and choriocarcinomas.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: BDCA-3; CD141; CD141 antigen; DC141; Fetomodulin; sCD141; soluble CD141; Thrombomodulin; TM
Gene Aliases: AHUS6; BDCA3; CD141; THBD; THPH12; THRM; TM
UniProt ID: (Human) P07204
Entrez Gene ID: (Human) 7056
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support