Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MLAPRGAAVL LLHLVLQRWL AAGAQATPQV FDLLPSSSQR LNPGALLPVL TDPALNDLYV ISTFKLQTKS SATIFGLYSS TDNSKYFEFT VMGRLNKAIL RYLKNDGKVH LVVFNNLQLA DGRRHRILLR LSNLQRGAGS LELYLDCIQV DSVHNLPRAF AGPSQKPETI ELRTFQRKPQ DFLEELKLVV RGSLFQVASL QDCFLQQSEP LAATGTGDFN RQFLGQMTQL NQLLGEVKDL LRQQVKETSF LRNTIAECQA CGPLKFQSPT PSTVVPPAPP APPTRPPRRC DSNPCFRGVQ CTDSRDGFQC GPCPEGYTGN GITCIDVDEC KYHPCYPGVH CINLSPGFRC DACPVGFTGP MVQGVGISFA KSNKQVCTDI DECRNGACVP NSICVNTLGS YRCGPCKPGY TGDQIRGCKA ERNCRNPELN PCSVNAQCIE ERQGDVTCVC GVGWAGDGYI CGKDVDIDSY PDEELPCSAR NCKKDNCKYV PNSGQEDADR DGIGDACDED ADGDGILNEQ DNCVLIHNVD QRNSDKDIFG DACDNCLSVL NNDQKDTDGD GRGDACDDDM DGDGIKNILD NCPKFPNRDQ RDKDGDGVGD ACDSCPDVSN PNQSDVDNDL VGDSCDTNQD SDGDGHQDST DNCPTVINSA QLDTDKDGIG DECDDDDDND GIPDLVPPGP DNCRLVPNPA QEDSNSDGVG DICESDFDQD QVIDRIDVCP ENAEVTLTDF RAYQTVVLDP EGDAQIDPNW VVLNQGMEIV QTMNSDPGLA VGYTAFNGVD FEGTFHVNTQ TDDDYAGFIF GYQDSSSFYV VMWKQTEQTY WQATPFRAVA EPGIQLKAVK SKTGPGEHLR NSLWHTGDTS DQVRLLWKDS RNVGWKDKVS YRWFLQHRPQ VGYIRVRFYE GSELVADSGV TIDTTMRGGR LGVFCFSQEN IIWSNLKYRC NDTIPEDFQE FQTQNFDRFD N
Thrombospondin-4 (THBS4) belongs to the thrombospondin protein family, a family of adhesive calcium-binding glycoproteins that mediate cell-to-cell and cell-to-matrix interactions. Studies suggest that this protein may be involved in local signaling in the developing and adult nervous system. THBS4 is widely expressed and is abundant in atherosclerotic lesions and in areas prone to the development of lesions. It has been suggested to play an important role in the recruitment of macrophages and the local regulation of inflammation associated with atherogenesis. THBS4 has also been suggested as a putative tumor-suppressor protein in colorectal cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Thrombospondin-4
Gene Aliases: THBS4; TSP-4; TSP4
UniProt ID: (Human) P35443
Entrez Gene ID: (Human) 7060
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support