Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MIRRCFQVAA RLGHHRGLLE APRILPRLNP ASAFGSSTDS MFSRFLPEKT DLKDYALPNA SWCSDMLSLY QEFLEKTKSS GWIKLPSFKS NRDHIRGLKL PSGLAVSSDK GDCRIFTRCI QVEGQGFEYV IFFQPTQKKS VCLFQPGSYL EGPPGFAHGG SLAAMMDETF SKTAFLAGEG LFTLSLNIRF KNLIPVDSLV VMDVEVDKIE DQKLYMSCIA HSRDQQTVYA KSSGVFLQLQ LEEESPQ
THEM5 (thioesterase superfamily member 5) is a 247 amino acid protein that belongs to the thioesterase superfamily. The gene that encodes THEM5 contains nearly 8,000 bases and maps to human chromosome 1q21.3. Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. The mechanism of rapidly enhanced aging is unclear and is a topic of continuing exploration. The MUTYH gene is located on chromosome 1 and is partially responsible for familial adenomatous polyposis. Stickler syndrome, Parkinsons, Gaucher disease and Usher syndrome are also associated with chromosome 1. Has acyl-CoA thioesterase activity towards long-chain (C16 and C18) fatty acyl-CoA substrates, with a preference for linoleyl-CoA and other unsaturated long-chain fatty acid-CoA esters. Plays an important role in mitochondrial fatty acid metabolism, and in remodeling of the mitochondrial lipid cardiolipin. Required for normal mitochondrial function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acyl-CoA thioesterase THEM5; Acyl-coenzyme A thioesterase 15; Acyl-coenzyme A thioesterase THEM5; Thioesterase superfamily member 5
Gene Aliases: ACOT15; THEM5
UniProt ID: (Human) Q8N1Q8
Entrez Gene ID: (Human) 284486
Molecular Function:
esterase
hydrolase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support