Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Bovine
THOC7 gene ontology annotations related to this gene include mRNA processing.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ23445; fSAP24; Functional spliceosome-associated protein 24; hTREX30; Ngg1 interacting factor 3 like 1 binding protein 1; Ngg1-interacting factor 3-like protein 1-binding protein 1; NIF3L1-binding protein 1; THO complex 7 homolog; THO complex subunit 7 homolog
Gene Aliases: fSAP24; hTREX30; NIF3L1BP1; THOC7
UniProt ID: (Human) Q6I9Y2
Entrez Gene ID: (Human) 80145
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support