Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
A 1:500 dilution of MA5-45921 was sufficient for detection of Pan-Thyroid hormone receptor in 10 µg of Hep G2 Human Hepatoblastoma Cell lysate by ECL immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody.
Detects 70 kDa.
The protein encoded by this gene is a nuclear hormone receptor for triiodothyronine. It is one of the several receptors for thyroid hormone, and has been shown to mediate the biological activities of thyroid hormone. Knockout studies in mice suggest that the different receptors, while having certain extent of redundancy, may mediate different functions of thyroid hormone. Mutations in this gene are known to be a cause of generalized thyroid hormone resistance (GTHR), a syndrome characterized by goiter and high levels of circulating thyroid hormone (T3-T4), with normal or slightly elevated thyroid stimulating hormone (TSH). Several alternatively spliced transcript variants encoding the same protein have been observed for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 1; c-erb-A thyroid hormone receptor; c-erbA-1; c-erbA-2; C-erbA-alpha; c-erbA-alpha2; c-erbA-beta; c-erbAalpha; c-erbAalpha2; EAR-7; ERBA-related 7; Nuclear receptor subfamily 1 group A member 1; Nuclear receptor subfamily 1 group A member 2; oncogene ERBA2; THR; thyroid hormone nuclear receptor beta variant 1; Thyroid hormone receptor alpha; thyroid hormone receptor alpha 1; thyroid hormone receptor alpha 1 (avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 1 formerly ERBA1); Thyroid hormone receptor beta; thyroid hormone receptor beta 3; thyroid hormone receptor beta2delta; thyroid hormone receptor, alpha (erythroblastic leukemia viral (v-erb-a) oncogene homolog, avian); thyroid hormone receptor, beta (avian erythroblastic leukemia viral (v-erb-a) oncogene homolog 2); thyroid hormone receptor, beta (erythroblastic leukemia viral (v-erb-a) oncogene homolog 2, avian); thyroid normone nuclear receptor alpha variant 1; TR alpha 1; TR alpha 2; TR beta; TRbeta2Delta; triiodothyronine receptor; V-erbA-related protein 7
Gene Aliases: 6430529J03Rik; AR7; AW259572; c-ERBA-1; C-ERBA-2; C-erba-alpha; C-ERBA-BETA; c-erbAbeta; CHNG6; EAR7; ERB-T-1; ERBA; ERBA1; ERBA2; GRTH; NR1A1; NR1A2; PRTH; RATT3REC; Rvr; T3R[a]; T3R[b]; T3Ralpha; T3Rbeta; T3rec; THR1; THRA; THRA1; THRA2; THRB; THRB1; THRB2; TRbeta
UniProt ID: (Human) P10827, (Human) P10828, (Mouse) P63058, (Rat) P63059, (Mouse) P37242
Entrez Gene ID: (Human) 7067, (Human) 7068, (Mouse) 21833, (Rat) 81812, (Mouse) 21834, (Rat) 24831
Molecular Function:
C4 zinc finger nuclear receptor
DNA-binding transcription factor
zinc finger transcription factor
gene-specific transcriptional regulator
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support