Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a component of the mediator complex (also known as TRAP, SMCC, DRIP, or ARC), a transcriptional coactivator complex thought to be required for the expression of almost all genes. The mediator complex is recruited by transcriptional activators or nuclear receptors to induce gene expression, possibly by interacting with RNA polymerase II and promoting the formation of a transcriptional pre-initiation complex. The product of this gene is proposed to form a sub-complex with MED12, cyclin C, and CDK8 that can negatively regulate transactivation by mediator. [provided by RefSeq].
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activator-recruited cofactor 250 kDa component; ARC250; DRIP250; Mediator complex subunit 13; Mediator of RNA polymerase II transcription subunit 13; mediator of RNA polymerase II transcription, subunit 13 homolog; thyroid hormone receptor associated protein 1; Thyroid hormone receptor-associated protein 1; Thyroid hormone receptor-associated protein complex 240 kDa component; thyroid hormone receptor-associated protein complex component TRAP240; thyroid hormone receptor-associated protein, 240 kDa subunit; Trap240; Vitamin D3 receptor-interacting protein complex component DRIP250
Gene Aliases: ARC250; DRIP250; HSPC221; KIAA0593; MED13; THRAP1; TRAP240
UniProt ID: (Human) Q9UHV7
Entrez Gene ID: (Human) 9969
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support