Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GQEVKIDQR NVKKEFTSHA LDSHILDYYE NPAIKEDVST LIGDDLASCK DETDESSKEE TEPQVLKFRV TCNRAGEKHC FTSNEAARDF GGAVQDYFKW KADMTNFDVE VLLNIHDNEV IVGIALTEES LHRRNITHFG PTTLRSTLAY GMLRLCDPLP YDIIVDPMCG TGAIPIEGAT EWSDCFHIAG DNNPLAVNRA ANNIASLLTK SQIKEGKPSW GLPIDAVQWD ICNLPLRTGS VDIIVTDLPF GKRMGSKKRN WNLYPACLRE MSRVCTPTTG RAVLLTQDTK CFTKALSGMR HVWRKVDTVW VNVGGLRAAV YVLIRTPQAF VHPSEQDGER GTLWQCKE (161-507 aa encoded by BC001622)
THUMPD3 a protein of unknown function. Its domain structure suggests that it is a methylase. It contains a THUMP domain (an RNA-binding domain) and a UPF0020 domain (putative methylase).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gene trap ROSA 26 antisense, Philippe Soriano; GtROSA26asSor; thum3; THUMP domain-containing protein 3; tRNA (guanine(6)-N2)-methyltransferase THUMP3
Gene Aliases: AW556087; Gt(ROSA)26asSor; Gtrosa26as; RGD1560852; THUMPD3
UniProt ID: (Human) Q9BV44, (Mouse) P97770
Entrez Gene ID: (Human) 25917, (Rat) 500288, (Mouse) 14911
Molecular Function:
RNA methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support