Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
THUMPD3 a protein of unknown function. Its domain structure suggests that it is a methylase. It contains a THUMP domain (an RNA-binding domain) and a UPF0020 domain (putative methylase).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gene trap ROSA 26 antisense, Philippe Soriano; GtROSA26asSor; thum3; THUMP domain-containing protein 3; tRNA (guanine(6)-N2)-methyltransferase THUMP3
Gene Aliases: AW556087; Gt(ROSA)26asSor; Gtrosa26as; THUMPD3
UniProt ID: (Human) Q9BV44, (Mouse) P97770
Entrez Gene ID: (Human) 25917, (Mouse) 14911
Molecular Function:
RNA methyltransferase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support