Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Positive control: NIH/3T3 cell lysate, MCF-7 cell lysate, RAW264.7 cell lysate, A549 cell lysate, PC-12 cell lysate, rat testis tissue, human kidney tissue, mouse colon tissue.
Predicted band size: 22 kDa
Subcellular Location: Mitochondrion inner membrane.
Essential component of the TIM23 complex, a complex that mediates the translocation of transit peptide-containing proteins across the mitochondrial inner membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial import inner membrane translocase subunit Tim23; translocase of inner mitochondrial membrane 23 homolog; translocase of the inner mitochondrial membrane
Gene Aliases: TIM23; TIMM23
UniProt ID: (Human) O14925, (Rat) O35093
Entrez Gene ID: (Human) 100287932, (Rat) 54312, (Mouse) 53600
Molecular Function:
amino acid transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support