Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Sandwich ELISA (Recombinant protein).
Immunogen sequence: KMYTGLCNFV ERWDQLTLSQ RKGLNYRYHL GCNCKIKSCY YLPCFVTSKN ECLWTDMLSN FGYPGYQSKH YACIRQKGGY CSWYRGWAPP DKSIINATDP
Tissue inhibitor of metalloproteinase 3 (TIMP3) is a member of the tissue inhibitor of metalloproteinases gene family. It functions to inhibit the activity of matrix metalloproteinases (MMP) which degrade components of the extracellular matrix. TIMP3 is expressed upon mitogenic stimulation. It binds irreversibly to zinc-dependent MMPs and inactivates them by binding to their catalytic zinc cofactor. Other functions of TIMP3 include nervous system development, tissue regeneration, and visual perception. In humans, the gene encoding TIMP3 is located on chromosome 22.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Metalloproteinase inhibitor 3; MIG-5 protein; Protein MIG-5; TIMP 3; TIMP-3; Tissue inhibitor of metalloproteinases 3
Gene Aliases: HSMRK222; K222; K222TA2; SFD; TIMP3
UniProt ID: (Human) P35625
Entrez Gene ID: (Human) 7078
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support