Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MADAEARAEF PEEARPDRGT LQVFQDMASR LRIHSIRATC STSSGHPTSC SSSSEIMSVL FFYIMRYKQS DPENPDNDRF VLAKRLSFVD VATGWLGQGL GVACGMAYTG KYFDRASYRV FCLMSDGESS EGSVWEAMAF ASYYSLDNLV ATFDVNRLGH SGALPAEHCI NIYQRRCEAF GWNTYVVDGR DVEALCQVFW QASQVKHKPT AVVAKTFKGR GTPSIEDAES WHAKPMPRER ADAIIKLIES QIQTSRNLDP QPPIEDSPEV NITDVRMTSP PDYRVGDKIA TRKACGLALA
Strong TKTL1 protein expression has been correlated with a certain type of glucose metabolism (aerobic glycolysis; Warburg effect) and to cells which are affected by chronic complications of diabetic patients. In colon and urothelial carcinomas, expression at the protein level is correlated with invasiveness of tumors and poor patient survival.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TK 2; Transketolase 2; transketolase-2; transketolase-like 1; Transketolase-like protein 1; Transketolase-related protein
Gene Aliases: TKR; TKT2; TKTL1
UniProt ID: (Human) P51854
Entrez Gene ID: (Human) 8277
Molecular Function:
transketolase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support