Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Plays an essential role in total transketolase activity and cell proliferation in cancer cells; after transfection with anti-TKTL1 siRNA, total transketolase activity dramatically decreases and proliferation was significantly inhibited in cancer cells. Plays a pivotal role in carcinogenesis. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: testis tissue sperm-binding protein Li 39a; transketolase-like 2; Transketolase-like protein 2
Gene Aliases: 4933401I19Rik; RGD1304767; TKTL2
UniProt ID: (Human) Q9H0I9, (Mouse) Q9D4D4
Entrez Gene ID: (Human) 84076, (Rat) 290685, (Mouse) 74419
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support