Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SALSVVQNLE KDLQEVKAAA RDGKLKPLPG ETMEKCTQDL GNSTKAVSSA IAQLLGEVAQ GNENYAGIAA RDVAGGLRSL AQAARGVAAL TSDPAVQA
Talin, a multifunctional constituent of cell-substratum attachment sites, is a high molecular weight protein (225-235 kDa) found in variety of tissues and cell types. It is localized at a subset of adherens junctions, specialized cell-cell and cell-matrix associations that are characterized by the presence of filamentous actin at the cytoplasmic face of the junctional complex. In cultured cells, talin is absent from cell-cell junctions and found predominantly at adhesion plaques and in fibrillar streaks underlying cell surface fibronectin. Talin interacts with at least two other proteins that are localized at adhesion plaques, vinculin and integrin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Talin-1
Gene Aliases: ILWEQ; KIAA1027; TLN; TLN1
UniProt ID: (Human) Q9Y490
Entrez Gene ID: (Human) 7094
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support