Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: DAPELPEERE LMTNCSNMSL RKVPADLTPA TTTLDLSYNL LFQLQSSDFH SVSKLRVLIL CHNRIQQLDL KTFEFNKELR YLDLSNNRLK SVTWYLLAGL RYLDLSFNDF DTMPICEEAG NMSHLEILGL SGAKIQKSDF QKIAHLHLNT VFLGFRTLPH YEEGSLPILN TTKLHIVLPM DTNFWVLLRD GIKTSKILEM TNIDGKSQFV SYEMQRNLSL ENAKTSVLLL NKVDLLWDDL FLILQFVWHT SVEHF
The protein encoded by this gene is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. This gene is most highly expressed in lymphoid tissues such as spleen, lymph node, thymus, and tonsil. Its exact function is not known. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD290; MGC104967; MGC126398; MGC126399; Toll-like receptor 10
Gene Aliases: CD290; TLR10; UNQ315/PRO358
UniProt ID: (Human) Q32MI8
Entrez Gene ID: (Human) 81793, (Rat) 305355
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support