Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 7
Positive Samples: A-549, LO2, U-87MG; Cellular Location: Early endosome, Endoplasmic reticulum membrane, Endosome membrane, Single-pass type I membrane protein
Immunogen sequence: SPFQPLRNLT ILDLSNNNIA NINDDMLEGL EKLEILDLQH NNLARLWKHA NPGGPIYFLK GLSHLHILNL ESNGFDEIPV EVFKDLFELK IIDLGLNNLN TLPASVFNNQ VSLKSLNLQK NLITSVEKKV FGPAFRNLTE LDMRFNPFDC TCESIAWFVN WINETHTNIP ELSSHYLCNT PPHYHGFPVR LFDTSSCKDS A
TLR3 (Toll-like receptor 3, CD283) is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from drosophila to humans and share structural and functional similarities. TLRs recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. TLR3 is most abundantly expressed in placenta and pancreas, and is restricted to the dendritic subpopulation of the leukocytes. TLR3 recognizes dsRNA associated with viral infection, and induces the activation of NF-kappaB and the production of type I interferons. Further, TLR3 may play a role in host defense against viruses, and the use of alternative polyadenylation sites to generate different length transcripts has been noted for the TLR3 gene. Like its counterparts in Drosophila, TLRs signal through adaptor molecules and could constitute an important and unrecognized component of innate immunity in humans. The TLR family is a phylogenetically conserved mediator of innate immunity that is essential for microbial recognition. TLRs characterized so far activate the MyD88/interleukin-1 receptor-associated kinase (IRAK) signaling pathway. TLR3 is the only human TLR which does not utilize MyD88 as an adaptor molecule instead utilizing TRIF in a signaling path which results in downstream activation of IRF-3 and NF-kB. TLR3 activation leads not only to Type I interferon induction but also to other inflammatory cytokines which result in DC maturation. TLR3 is known to recognize viral double-stranded (ds) RNA, a molecular pattern associated with viral infection. Recently it has been shown to recognize viruses such as Influenza A and West Nile Virus and can mediate entry of at least West Nile Virus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD283; TLR 3; Toll-like receptor 3
Gene Aliases: AI957183; CD283; IIAE2; TLR3
UniProt ID: (Human) O15455, (Mouse) Q99MB1
Entrez Gene ID: (Human) 7098, (Mouse) 142980, (Rat) 364594
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support