Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Reconstitute with 1 mL of distilled water. After reconstitution, store product undiluted at 4ºC in the dark.
For FACS analysis, use 10 µL of the suggested working dilution to label 1x10^6 cells in 100 µL.
TLR4 is member of the Toll like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. This receptor is most abundantly expressed in placenta, and in myelomonocytic subpopulation of the leukocytes. Mammalian cells respond to LPS by activating TLR4. TLR4 belongs to the multi-protein complex of lipopolysaccharide (LPS) receptor, containing CD14, LY96 and TLR4, and is involved in signal transduction events induced by lipopolysaccharide (LPS) found in most gram-negative bacteria. TLR4 aids in the recognition of pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. Mutations in the TLR4 gene have been associated with differences in LPS responsiveness. Also, several transcript variants of the TLR4 gene have been found, but the protein coding potential of most of them is uncertain. TLR4 is expressed by peripheral blood monocytes and a small population of B-cells and is also expressed in human placenta. Studies with TLR4-deficient mice indicate that the main ligand for TLR is lipopolysaccharide. Consequently, these mice also showed increased susceptibility to Gram-negative sepsis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD284; homolog of Drosophila toll; hToll; TLR4; Toll-like receptor 4; Toll-like receptor4 protein; toll/interleukin-l receptor (TIR) domain
Gene Aliases: ARMD10; CD284; EGK_07381; hToll; TLR-4; TLR4; TOLL
UniProt ID: (Human) O00206
Entrez Gene ID: (Rhesus monkey) 574360, (Pig) 399541, (Human) 7099, (Dog) 403417, (Guinea pig) 100724803
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support