Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MD-2 is a member of the Toll/interleukin-1 receptor (TIR) family, a group of proteins that include the Toll-like receptors (TLRs). TLRs are signaling molecules that recognize different pathogen-associated molecular patterns (PAMPs) and serve as an important link between the innate and adaptive immune responses. TLR4, the major signaling receptor for lipopolysaccharide (LPS), requires the binding of MD-2 to its extracellular region for maximal response to LPS. The specificity of this response is determined by the species of MD-2; e.g., human MD-2 transfected into mouse cells can cause mouse TLR4 to react to LPS analogs that are normally antagonistic to human but not mouse TLR4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ARMD10; CD284; ESOP-1; lipopolysaccharide response; Ly-96; Lymphocyte antigen 96; myeloid differentiation factor-2; myeloid differentiation protein-2; Protein MD-2; TLR-4; TOLL; Toll-like receptor 4
Gene Aliases: ESOP-1; Esop1; Lps; Ly87; Ly96; MD-2; MD2; Ran/M1; Rasl2-8; Tlr4
UniProt ID: (Mouse) Q9JHF9, (Mouse) Q9QUK6
Entrez Gene ID: (Mouse) 17087, (Mouse) 21898
Molecular Function:
scaffold/adaptor protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support