Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TLR6 (Toll-like receptor 6) is a member of the toll-like receptor family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved and share structural and functional similarities. TLRs recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. TLR6 functionally interacts with TLR2 to mediate cellular response to bacterial lipoproteins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD antigen 286; CD286; Toll-like receptor 6
Gene Aliases: CD286; TLR-6; TLR6
UniProt ID: (Human) Q9Y2C9, (Mouse) Q9EPW9
Entrez Gene ID: (Pig) 396621, (Cat) 493662, (Human) 10333, (Mouse) 21899, (Rat) 305353
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support