Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: ARWFPKTLPC DVTLDVPKNH VIVDCTDKHL TEIPGGIPTN TTNLTLTINH IPDISPASFH RLDHLVEIDF RCNCVPIPLG SKNNMCIKRL QIKPRSFSGL
TLR7 is a 10 amino acid long highly conserved membrane protein belonging to the Toll-like receptor (TLR) family with 27 LRR (leucine-rich) repeats and a TIR domain. TLR7 plays a fundamental role in pathogen recognition and activation of innate immunity. TLR7 is activated by infections with single-stranded RNA viruses, including influenza virus and vesicular stomatitis virus (VSV). TLR7 is predominantly expressed in lung, placenta, and spleen, and lies in close proximity to another family member, TLR8. TLR7 also acts via MyD88 and TRAF6, leading to NF-kappaB activation, cytokine secretion and the inflammatory response. TLR7 recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RP23-139P21.3; Toll-like receptor 7; toll-like receptor 7-like
Gene Aliases: TLR7; TLR7-like; UNQ248/PRO285
UniProt ID: (Human) Q9NYK1
Entrez Gene ID: (Human) 51284
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support