Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 6
Positive Samples: Mouse liver; Cellular Location: Membrane, Single-pass type I membrane protein
Immunogen sequence: HHLFYWDVWF IYNVCLAKVK GYRSLSTSQT FYDAYISYDT KDASVTDWVI NELRYHLEES RDKNVLLCLE ERDWDPGLAI IDNLMQSINQ SKKTVFVLTK KYAKSWNFKT AFYLALQRLM DENMDVIIFI LLEPVLQHSQ YLRLRQRICK SSILQWPDNP KAEGLFWQTL RNVVLTENDS RYNNMYVDSI KQY
The TLR8 protein is a member of the Toll-like receptor (TLR) family which plays a fundamental role in pathogen recognition and activation of innate immunity. TLRs are highly conserved from Drosophila to humans and share structural and functional similarities. They recognize pathogen-associated molecular patterns (PAMPs) that are expressed on infectious agents, and mediate the production of cytokines necessary for the development of effective immunity. The various TLRs exhibit different patterns of expression. The TLR8 gene is predominantly expressed in lung and peripheral blood leukocytes, and lies in close proximity to another family member, TLR7, on chromosome X.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD antigen 288; CD288; Toll-like receptor 8
Gene Aliases: CD288; TLR8; UNQ249/PRO286
UniProt ID: (Human) Q9NR97, (Mouse) P58682
Entrez Gene ID: (Human) 51311, (Mouse) 170744, (Rat) 684440
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support