Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: ETLQHVPSDH TNETSNSTVK PPTSVASDSS NTTVTTMKPT AASNTTTPGM VSTNMTSTTL KSTPKTTSVS QNTSQISTST MTVTHNSSVT SAASSVTITT
Transmembrane protein 123, or Tmem123, is a single-pass type I membrane protein and belongs to the CD164 family. Tmem123 acts as a novel marker for DC maturation. DCs are also known as professional antigen-presenting cells. It is reported that the Tmem123 molecule may be closely associated with the cell surface expression of CD40. It was also reported previously that Tmem123 protein may also be responsible for oncotic cell death, which is characterized by cell swelling, organelle swelling, vacuolization, increased membrane permeability and cell adhesion.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KCT-3; keratinocytes associated transmembrane protein 3; Keratinocytes-associated transmembrane protein 3; Porimin; pro oncosis receptor inducing membrane injury; Pro-oncosis receptor inducing membrane injury; serine/threonine-rich receptor; Transmembrane protein 123
Gene Aliases: KCT3; PORIMIN; PORMIN; PSEC0111; TMEM123; UNQ641/PRO1271
UniProt ID: (Human) Q8N131
Entrez Gene ID: (Human) 114908
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support