Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
A suggested positive control is rat brain tissue lysate.
PA5-20842 can be used with blocking peptide PEP-0956.
Calcium-activated chloride channels (CaCC) are present in many cell types and mediate physiological functions such as epithelial secretion, sensory signal transduction, and smooth muscle contraction. Subunits of these CaCC’s include the transmembrane proteins TMEM16A and TMEM16B. TMEM16B is predicted to have eight transmembrane domains with both the amino and carboxy termini in the cytoplasm and is expressed in several tissues including olfactory sensory neurons as well as photoreceptors in mammalian retina. Like TMEM16A, TMEM16B is thought to form at least part of CaCC’s but has different biophysical characteristics such as voltage dependence and unitary conductance.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ano2; anoctamin 2 isoform Adelta4; anoctamin 2 isoform Bdelta4; anoctamin 2, calcium activated chloride channel; anoctamin 2-like; Anoctamin-2; Transmembrane protein 16B; transmembrane protein 16B (eight membrane-spanning domains)
Gene Aliases: ANO2; BC033409; C12orf3; TMEM16B
UniProt ID: (Human) Q9NQ90, (Mouse) Q8CFW1
Entrez Gene ID: (Human) 57101, (Mouse) 243634, (Rat) 100361584
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support