Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This antibody is specific to TMEM181.
Mediates action of cytolethal distending toxins (CDT), which are secreted by many pathogenic bacteria. Expression level of TMEM181 is rate-limiting for intoxication.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 178; Transmembrane protein 181
Gene Aliases: 5930418K15Rik; C76977; GPR178; KIAA1423; mKIAA1423; TMEM181
UniProt ID: (Human) Q9P2C4
Entrez Gene ID: (Human) 57583, (Mouse) 77106
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support