Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VTACAAAGV LLYRIARRMK PTHTMVNCWF CNQDTLVPYG NRNCWDCPHC EQYNGFQENG DYNKPIPAQY LEHLNHVVSS APSLRDPSQP QQWVSSQVLL CKRCNHHQTT KIKQLAAFAP REEGRYDEEV EVYRHHLEQM YKLCRPCQAA VEYYIKHQNR QLRALLLSHQ FKRREADQTH AQNFSSAVKS PVQVILLR (22-218 aa encoded by BC137291 )
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: RP13-15M17.2; Spindle-associated membrane protein 1; Transmembrane protein 201
Gene Aliases: AV028368; D4Ertd429e; NET5; SAMP1; TMEM201
UniProt ID: (Human) Q5SNT2, (Mouse) A2A8U2
Entrez Gene ID: (Human) 199953, (Mouse) 230917
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support