Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: VRLSIRTAL GDKAYAWDTN EEYLFKAMVA FSMRKVPNRE ATEISHVLLC NVTQRVSFWF VVTDPSKNHT LPAVEVQSAI RMNKNRINNA FFLNDQTLEF LKIPSTLAPP MDPSVPIW (27-143 aa encoded by BC050606 )
TMEM27 (Transmembrane protein 27) is also named as collectrin and Belongs to the TMEM27 family. It is a stimulator of beta cell replication and a membrane protein cleaved and shed by pancreatic beta cells that has been proposed as a beta cell mass biomarker. Western blot analysis of mouse collecting duct cell lines detected Tmem27 at an apparent molecular mass of 32 kDa. TMEM27 is an N-glycosylated protein and forms dimers, with MW of 64-75 kDa.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Collectrin; kidney-specific membrane protein; Transmembrane protein 27
Gene Aliases: 0610008J07Rik; CLTRN; NX-17; NX17; TMEM27; UNQ679/PRO1312
UniProt ID: (Human) Q9HBJ8, (Mouse) Q9ESG4
Entrez Gene ID: (Human) 57393, (Mouse) 57394
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support