Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: GSQTSSWEKL EKNLSDRLGE SQTLLAAAVE RERAATEELL ANKIQMSSME SQNSLLRQEN SRFQAQLESE KNRLCKLEDE NNRYQVELEN LKDEYVRTLE ETRKEKTLLN SQLEMERMKV EQERKKAIFT QETIKEKERK PFSVSSTPTM SRSSSISGVD MAGLQTSFLS QDESHDHSFG PMPISANGSN LYDAVRMGAG SSIIENLQSQ LKLREGEITH LQLEIGNLEK TRSIMAEELV KLTNQNDELE EKVKEIPKLR TQLRDLDQRY NTILQMYGEK AEEAEELRLD LEDVKNMYKT QIDELLRQSL S
Adapter protein that may provide indirect link between the endocytic membrane traffic and the actin assembly machinery. May regulate the formation of clathrin-coated vesicles. Isoform 1 could be involved in brain-specific synaptic vesicle recycling.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Androgen receptor coactivator 160 kDa protein; Androgen receptor-associated protein of 160 kDa; TATA element modulatory factor; TMF
Gene Aliases: 7030402D04Rik; ARA160; Gm153; TMF; TMF1; Tmf1`
UniProt ID: (Human) P82094, (Mouse) B9EKI3
Entrez Gene ID: (Human) 7110, (Rat) 114206, (Mouse) 232286
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support