Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Relative expression to ensure that the antibody binds to the antigen stated.
Immunogen sequence: LEKEALEHKD REDYVPYTGE KKGKIFIPKQ KPVQTFTEEK VSLDPELEEA LTSASDTELC DLAAILGMHN LITNTKFCNI MGSSNGVDQE HFSNVVKGEK ILPVFDEPPN PTNVEESLKR TKENDAHLVE VNLNNIKNIP IPTLKD
Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.
Blocks the elongation and depolymerization of the actin filaments at the pointed end. The Tmod/TM complex contributes to the formation of the short actin protofilament, which in turn defines the geometry of the membrane skeleton.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: tropomodulin 3 (ubiquitous); tropomodulin 3; ubiquitous tropomodulin; Tropomodulin-3; U-Tmod; Ubiquitous tropomodulin
Gene Aliases: TMOD3; U-Tmod; UTMOD
UniProt ID: (Human) Q9NYL9, (Mouse) Q9JHJ0
Entrez Gene ID: (Human) 29766, (Mouse) 50875, (Rat) 300838
Molecular Function:
actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support