Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: KGPNCSEPEC PGNCHLRGRC IDGQCICDDG FTGEDCSQLA CPSDCNDQGK CVNGVCICFE GYAGADCSRE ICPVPCSEEH GTCVDGLCVC HDGFAGDDCN KPLCLNNCYN
Tenascin is a high molecular weight, multifunctional, extracellular matrix glycoprotein expressed in association with mesenchymal-epithelial interactions during development and in the neovasculature and stroma of undifferentiated tumors (Erickson and Bourdon 1989). It has been described under a variety of names: cytotactin, hexabrachion protein, J1, myotendinous antigen (MI) and glioma mesenchymal extracellular matrix (GMEM). The tenascin molecule is a disulfide-linked hexamer. The expression of tenascin is associated with development and growth, both normal and pathological, whereas the distribution in normal adult tissue is restricted.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cytotactin; deafness, autosomal dominant 56; Glioma-associated-extracellular matrix antigen; GMEM; GP 150-225; Hexabrachion; hexabrachion (tenascin); JI; MGC144208; MGC144209; MGC167029; Myotendinous antigen; Neuronectin; Tenascin; Tenascin-C; tenascin-C additional domain 1; tenascin-C isoform 14/AD1/16; TN; TN-C
Gene Aliases: 150-225; DFNA56; GMEM; GP; HXB; JI; TN; TN-C; TNC
UniProt ID: (Human) P24821
Entrez Gene ID: (Human) 3371
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support