Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MAPPPARVHL GAFLAVTPNP GSAASGTEAA AATPSKVWGS SAGRIEPRGG GRGALPTSMG QHGPSARARA GRAPGPRPAR EASPRLRVHK TFKFVVVGVL LQVVPSSAAT IKLHDQSIGT QQWEHSPLGE LCPPGSHRSE HPGACNRCTE GVGYTNASNN LFACLPCTAC KSDEEERSPC TTTRNTACQC KPGTFRNDNS AEMCRKCSRG CPRGMVKVKD CTPWSDIECV HKESGNGHNI WVILVVTLVV PLLLVAVLIV CCCIGSGCGG DPKCMDRVCF WRLGLLRGPG AEDNAHNEIL SNADSLSTFV SEQQMESQEP ADLTGVTVQS PGEAQCLLGP AEAEGSQRRR LLVPANGADP TETLMLFFDK FANIVPFDSW DQLMRQLDLT KNEIDVVRAG TAGPGDALYA MLMKWVNKTG RNASIHTLLD ALERMEERHA KEKIQDLLVD SGKFIYLEDG TGSAVSLE
TRAIL-R1 (CD261, DR4) is a type I transmembrane protein, also called TRAIL receptor 1. The ligand for this DR4 death receptor has been identified and termed TRAIL, which is a member of the TNF family. DR4, as many other receptors (Fas, TNFR1, etc.), mediates apoptosis and NF kappaB activation in many cells and tissues. Apoptosis, a programmed cell death, is a operating process in normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by coupled of certain cytokines (TNF family - TNF, Fas ligand) and their death domain containing receptors (TNFR1, Fas receptor).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CD261; cytotoxic TRAIL receptor; Death receptor 4; MGC9365; sCD261; soluble CD261; soluble TRAIL; sTRAIL; TNF-related apoptosis-inducing ligand receptor 1; TRAIL R1; TRAIL receptor 1; TRAIL-R1; Tumor necrosis factor receptor superfamily member 10A; tumor necrosis factor receptor superfamily member 10a variant 2; tumor necrosis factor receptor superfamily, member 10a
Gene Aliases: APO2; CD261; DR4; TNFRSF10A; TRAILR-1; TRAILR1
UniProt ID: (Human) O00220
Entrez Gene ID: (Human) 8797
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support