Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MARIPKTLKF VVVIVAVLLP VLAYSATTAR QEEVPQQTVA PQQQRHSFKG EECPAGSHRS EHTGACNPCT EGVDYTNASN NEPSCFPCTV CKSDQKHKSS CTMTRDTVCQ CKEGTFRNEN SPEMCRKCSR CPSGEVQVSN CTSWDDIQCV EEFGANATVE TPAAEETMNT SPGTPAPAAE ETMNTSPGTP APAAEETMTT SPGTPAPAAE ETMTTSPGTP APAAEETMTT SPGTPASSHY LSCTIVGIIV LIVLLIVFV
TRAIL-R3 (CD263) is a member of the TNF-receptor superfamily. This receptor contains an extracellular TRAIL-binding domain and a transmembrane domain, but no cytoplasmic death domain. This receptor is not capable of inducing apoptosis, and is thought to function as an antagonistic receptor that protects cells from TRAIL-induced apoptosis. This gene was found to be a p53-regulated DNA damage-inducible gene. The expression of this gene was detected in many normal tissues but not in most cancer cell lines, which may explain the specific sensitivity of cancer cells to the apoptosis-inducing activity of TRAIL.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Antagonist decoy receptor for TRAIL/Apo-2L; CD263; cytotoxic TRAIL receptor-3; DcR1; Decoy receptor 1; Decoy TRAIL receptor without death domain; Lymphocyte inhibitor of TRAIL; MGC149501; MGC149502; TNF-related apoptosis-inducing ligand receptor 3; TRAIL receptor 3; TRAIL receptor without an intracellular domain; Tumor necrosis factor receptor superfamily member 10C; tumor necrosis factor receptor superfamily, member 10c, decoy without an intracellular domain
Gene Aliases: CD263; DCR1; DCR1-TNFR; LIT; TNFRSF10C; TRAIL-R3; TRAILR3; TRID; UNQ321/PRO366
UniProt ID: (Human) O14798
Entrez Gene ID: (Human) 8794
Molecular Function:
transmembrane signal receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support