Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: ESGDCRQQEF RDRSGNCVPC NQCGPGMELS KECGFGYGED AQCVTCRLHR FKEDWGFQKC KPCLDCAVVN RFQKANCSAT SDAICGDCLP
TROY can mediate activation of c-Jun and NF-kappa-B. May promote caspase-independent cell death. Isoform 2 and isoform 3 may act as decoy receptors.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Toxicity and JNK inducer; TRADE; Tumor necrosis factor receptor superfamily member 19; tumor necrosis factor receptor superfamily, member 19
Gene Aliases: TAJ; TAJ-alpha; TNFRSF19; TRADE; TROY; UNQ1888/PRO4333
UniProt ID: (Human) Q9NS68
Entrez Gene ID: (Human) 55504
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support