Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MDDIYKAAV EQLTEEQKNE FKAAFDIFVL GAEDGCISTK ELGKVMRMLG QNPTPEELQE MIDEVDEDGS GTVDFDEFLV MMVRCMKDDS KGKSEEELSD LFRMFDKNAD GYIDLDELKI MLQATGETIT EDDIEELMKD GDKNNDGRID YDEFLEFMKG VE (1-161 aa encoded by BC030244)
Troponin (Tn) is the central regulatory protein of striated muscle contraction. Tn consists of three components: Tn-I which is the inhibitor of actomyosin ATPase, Tn-T which contains the binding site for tropomyosin and Tn-C. The binding of calcium to Tn-C abolishes the inhibitory action of Tn on actin filaments, and as a result of calcium-ion uptake by troponin-C, the elements of the muscle thin filament undergo a series of allosteric changes which allow the interaction of actin with myosin, the hydrolysis of ATP, and the generation of tension.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cardiac 3; cardiac troponin C; CMD1FF; CMD2A; CMH7; MGC116817; RCM1; slow twitch skeletal/cardiac muscle troponin C; TN-C; Tn1; Tni; TNN I3; TNNC 1; TNNI3; troponin C type 1 (slow); troponin C, cardiac/slow skeletal; Troponin C, slow skeletal and cardiac muscles; troponin C1, slow; Troponin I cardiac; TroponinI
Gene Aliases: AI874626; CMD1Z; CMH13; cTnC; cTnI; TN-C; TNC; Tncc; TNNC; TNNC1
UniProt ID: (Human) P63316, (Mouse) P19123
Entrez Gene ID: (Human) 7134, (Rat) 290561, (Mouse) 21924
Molecular Function:
actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support