Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSTSRKLKSH GMRRSKSRSP HKGVKRGGSK RKYRKGNLKS RKRGDDANRN YRSHL
TNP1 is a spermatid-specific product of the haploid genome which replaces histone and is itself replaced in the mature sperm by the protamines. Transition protein-1 is a spermatid-specific product of the haploid genome which replaces histone and is itself replaced in the mature sperm by the protamines (see PRM1, MIM 182880; PRM2, MIM 182890) (Luerssen et al., 1990 ). Sequence Note: removed 4 bases from the 5' end that did not align to the reference genome assembly.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Spermatid nuclear transition protein 1; STP-1; tnp; TP-1; transition protein 1 (during histone to protamine replacement)
Gene Aliases: TNP1; TP1
UniProt ID: (Human) P09430
Entrez Gene ID: (Human) 7141
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support