Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 3
Immunogen sequence: FTLPPLGGSL GAGTSTSRSS ERTPGAATAS ASGAAEDGAC GCLPNPGTFE ECHRKCKELF PIQM
Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 73%.
TOMM40 is the channel-forming subunit of the translocase of the mitochondrial outer membrane complex that is essential for protein import into mitochondria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Mitochondrial import receptor subunit TOM40 homolog; mitochondrial outer membrane protein; p38.5; PER EC1; Protein Haymaker; Translocase of outer membrane 40 kDa subunit homolog; translocase of outer mitochondrial membrane 40 homolog
Gene Aliases: C19orf1; D19S1177E; PER-EC1; PEREC1; TOM40; TOMM40
UniProt ID: (Human) O96008
Entrez Gene ID: (Human) 10452
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support