Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MGNVLAASS PPAGPPPPPA PALVGLPPPP PSPPGFTLPP LGGSLGAGTS TSRSSERTPG AATASASGAA EDGACGCLPN PGTFEECHRK CKELFPIQME GVKLTVNKGL SNHFQVNHTV ALSTIGESNY HFGVTYVGTK QLSPTEAFPV LVGDMDNSGS LNAQVIHQLG PGLRSKMAIQ TQQSKFVNWQ VDGEYRGSDF TAAVTLGNPD VLVGSGILVA HYLQSITPCL ALGGELVYHR RPGEEGTVMS LAGKYTLNNW LATVTLGQAG MHATYYHKAS DQLQVGVEFE ASTRMQDTSV SFGYQLDLPK ANLLFKGSVD SNWIVGATLE KKLPPLPLTL ALGAFLNHRK NKFQCGFGLT IG (1-361 aa encoded by BC017224)
TOMM40 is the channel-forming subunit of the translocase of the mitochondrial outer membrane complex that is essential for protein import into mitochondria.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: haymaker protein; Mitochondrial import receptor subunit TOM40 homolog; mitochondrial outer membrane protein; mitochondrial outer membrane protein 35; Mitochondrial outer membrane protein of 35 kDa; Mitochondrial outer membrane protein of 38 kDa; mitochondrial translocase of outer membrane protein; MOM35; OM38; outer membrane protein 38 kDa; p38.5; PER EC1; Protein Haymaker; Translocase of outer membrane 40 kDa subunit homolog; translocase of outer mitochondrial membrane 40 homolog
Gene Aliases: AW539759; C19orf1; D19S1177E; Mom35; OM38; PER-EC1; PEREC1; TOM40; Tom40a; TOMM40
UniProt ID: (Human) O96008, (Rat) Q75Q40, (Mouse) Q9QYA2
Entrez Gene ID: (Human) 10452, (Pig) 102165927, (Rat) 308416, (Mouse) 53333
Molecular Function:
primary active transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support