Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
May play a significant role in p53/TP53-mediating signaling pathway.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: P3H3; TP53 target 5; TP53-inducible gene 5 protein; TP53-target gene 5 protein
Gene Aliases: C20orf10; CLG01; RGD1561934; TP53TG5
UniProt ID: (Human) Q9Y2B4
Entrez Gene ID: (Human) 27296, (Rat) 499940
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support