Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: GLHLGVTPSV IRKRYNLTSQ DVGSGTSNNS QACAQFLEQY FHDSDLAQFM RLFGGNFAHQ ASVARVVGQQ GRGRAGIEAS LDVQYLMSAG ANISTWVYSS PGRHEGQEPF
This gene encodes a member of the sedolisin family of serine proteases. The protease functions in the lysosome to cleave N-terminal tripeptides from substrates, and has weaker endopeptidase activity. It is synthesized as a catalytically-inactive enzyme which is activated and auto-proteolyzed upon acidification. Mutations in this gene result in late-infantile neuronal ceroid lipofuscinosis, which is associated with the failure to degrade specific neuropeptides and a subunit of ATP synthase in the lysosome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cell growth-inhibiting gene 1 protein; CLN 2; GIG 1; growth-inhibiting protein 1; LPIC; lysosomal pepstatin insensitive protease; Lysosomal pepstatin-insensitive protease; MGC21297; TPP 1; TPP I; TPP-1; TPP-I; TPPI; Tripeptidyl aminopeptidase; tripeptidyl peptidase I; Tripeptidyl-peptidase 1; Tripeptidyl-peptidase I
Gene Aliases: CLN2; GIG1; LPIC; SCAR7; TPP-1; TPP1; UNQ267/PRO304
UniProt ID: (Human) O14773
Entrez Gene ID: (Human) 1200
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support