Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: LERLKDLPF IVSHRLSNTL SLDIHENHSF ALLGKKKSSN LTLPPKYNQP FFVTSLPDDK IPKGAGPGCY LAGSLTLSKT ELGKKADVIP VHYYLIPPPT KTKNGSKDKE KDSEKEKDLK EEFTEALRDL KIQWMTKLDS SDIYNELKET YPNYLPLYVA RLHQLDAEKE RMKRLNEIVD AANAVISHID QTALAVYIAM KTDPRPDAAT IKNDMDKQKS TLVDALCRKG CALADHLLHT QAQDGAISTD AEGKEEEGES PLDSLAETFW ETTKWTDLFD NKVLTFAYKH ALVNKMYGRG LKFATKLVEE KPTKENWKNC IQLMKLLGWT HCASFTENWL PIMYPPDYCV F (900-1249 aa encoded by BC039905)
This gene encodes a ubiquitously expressed transcription factor that controls cholesterol homeostasis by stimulating transcription of sterol-regulated genes. The encoded protein contains a basic helix-loop-helix-leucine zipper (bHLH-Zip) domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TPP-2; TPP-II; Tripeptidyl aminopeptidase; tripeptidyl peptidase II; Tripeptidyl-peptidase 2; Tripeptidyl-peptidase II
Gene Aliases: TPP-2; TPP2; TPPII
UniProt ID: (Human) P29144
Entrez Gene ID: (Human) 7174
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support