Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLSLLLLAL PVLASPAYAA PAPVQALQQA GIVGGQEAPR SKWPWQVSLR VRDRYWMHFC GGSLIHPQWV LTAAHCLGPD VKDLATLRVQ LREQHLYYQD QLLPVSRIIV HPQFYIIQTG ADIALLELEE PVNISSRVHT VMLPPASETF PPGMPCWVTG WGDVDNDEPL PPPFPLKQVK VPIMENHICD AKYHLGAYTG DDVRIIRDDM LCAGNTRRDS CQGDSGGPLV CKVNGTWLQA GVVSWDEGCA QPNRPGIYTR VTYYLDWIHH YVPKKP (1-275 aa encoded by B C028059)
Tryptase is a protein that belongs to the family of serine proteases and is primarily found in mast cells. It plays a crucial role in the immune response by cleaving and activating other proteins involved in inflammation and tissue repair. Tryptase is also involved in the regulation of blood pressure and has been implicated in various diseases, including asthma, allergies, and cardiovascular disorders. Its structure consists of four identical subunits that form a tetramer, and it is activated by proteolytic cleavage of its proenzyme form.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mast cell alpha II tryptase; mast cell beta I tryptase; TPSG1/TPSB2/TPSAB1/TPSD1; tryptase alpha II; Tryptase alpha-1; Tryptase alpha/beta-1; tryptase beta 1; tryptase beta I; tryptase beta-1; Tryptase I; Tryptase-1; tryptase-I; tryptase-III
Gene Aliases: TPS1; TPS2; TPSAB1; TPSB1
UniProt ID: (Human) Q15661
Entrez Gene ID: (Human) 7177
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support