Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-17190 targets TRAFD1 in indirect ELISA, FACS, ICC, IF, and WB applications and shows reactivity with Human samples.
The MA5-17190 immunogen is purified recombinant fragment of human TRAFD1 (amino acids: 401-582) expressed in E. Coli.
MA5-17190 detects TRAFD1 which has a predicted molecular weight of approximately 64.8kDa.
The innate immune system confers host defense against viral and microbial infection, and TRAFD1 is a negative feedback regulator that controls excessive immune responses.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLN29 gene product; Protein FLN29; TRAF-type zinc finger domain-containing protein 1
Gene Aliases: FLN29; TRAFD1
UniProt ID: (Human) O14545
Entrez Gene ID: (Human) 10906
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support