Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MAIFSVYVVN KAGGLIYQLD SYAPRAEAEK TFSYPLDLLL KLHDERVLVA FGQRDGIRVG HAVLAINGMD VNGRYTADGK EVLEYLGNPA NYPVSIRFGR PRLTSNEKLM LASMFHSLFA IGSQLSPEQG SSGIEMLETD TFKLHCYQTL TGIKFVVLAD PRQAGIDSLL RKIYEIYSDF ALKNPFYSLE MPIRCELFDQ NLKLALEVAE KAGTFGPGS
TRAPPC4 is part of the multisubunit TRAPP (transport protein particle) complex and interacts with SDC2. It may play a role in vesicular transport from endoplasmic reticulum to Golgi. TRAPP proteins are involved in tethering during vesicle transport.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hematopoietic stem/progenitor cell protein 172; Synbindin; syndecan binding protein 2; Trafficking protein particle complex subunit 4; TRS23 homolog
Gene Aliases: 1500017G03Rik; AI303617; CGI-104; HSPC172; PTD009; Sbd; SBDN; Sdcbp2; SYNBINDIN; TRAPPC4; TRS23
UniProt ID: (Human) Q9Y296, (Mouse) Q9ES56
Entrez Gene ID: (Human) 51399, (Mouse) 60409
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support