Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEPLRVLELY SGVGGMHHAL RESCIPAQVV AAIDVNTVAN EVYKYNFPHT QLLAKTIEGI TLEEFDRLSF DMILMSPPCQ PFTRIGRQGD MTDSRTNSFL YILDILPRLQ KLPKYILLEN VKGFEVSSTR DLLIQTIENC GFQYQEFLLS PTSLGIPNSR LRYFLIAKLQ SEPLPFQAPG QVLMEFPKIE SVHPQKYAMD VENKIQEKNV EPNISFDGSI QCSGKDAILF KLETAEEIHR KNQQDSDLSV KMLKDFLEDD TDVNQYLLPP KSLLRYALLL DIVQPTCRRS VCFTKGYGSY IEGTGSVLQT AEDVQVENIY KSLTNLSQEE QITKLLILKL RYFTPKEIAN LLGFPPEFGF PEKITVKQRY RLLGNSLNVH VVAKLIKILY E
CpG methylation is an epigenetic modification that is important for embryonic development, imprinting, and X-chromosome inactivation. Studies in mice have demonstrated that DNA methylation is required for mammalian development. This gene encodes a protein with similarity to DNA methyltransferases, but this protein does not display methyltransferase activity. The protein strongly binds DNA, suggesting that it may mark specific sequences in the genome. Alternative splicing results in multiple transcript variants encoding different isoforms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA (cytosine-5)-methyltransferase-like protein 2; DNA cytosine-5 methyltransferase 2; DNA methyltransferase homolog HsaIIP; DNA methyltransferase-2; DNA MTase homolog HsaIIP; Dnmt2; M.HsaIIP; PuMet; RP11-406H21.1; tRNA (cytosine(38)-C(5))-methyltransferase; tRNA (cytosine-5-)-methyltransferase
Gene Aliases: DMNT2; DNMT2; MHSAIIP; PUMET; RNMT1; TRDMT1
UniProt ID: (Human) O14717
Entrez Gene ID: (Human) 1787
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support