Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: PPPCESEIYC HGELLNQVQM AKLYQDDKQF VDMPLSIAPE QVLQTFTELS RDHNHSIPRE QLQAFVHEHF QAKGQELQPW TPADWKDSPQ FLQKISDAKL
This gene encodes an enzyme that hydrolyses trehalose, a disaccharide formed from two glucose molecules found mainly in fungi, plants, and insects. A partial duplication of this gene is located adjacent to this locus on chromosome 11.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Alpha,alpha-trehalase; Alpha,alpha-trehalose glucohydrolase; Trehalase; trehalase (brush-border membrane glycoprotein)
Gene Aliases: TRE; TREA; TREH
UniProt ID: (Human) O43280
Entrez Gene ID: (Human) 11181
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support