Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: AFCVDSITAL KALERASSPS EHGPRKSYSL GSIYTRLYGQ SPPDSHTAEG DVLALLSICQ WRPQALLRWV DAHARPFGTI RPMYGVTASA RTKPRPSAVT TTAHLATTRN TSPSLGESRG TKDLPPVKDP GALSREGLLA PLGLLAILTL AVATLYGLSL ATPGE
Trex1 (ATRIP) is the major human 3' to 5' exonuclease which is required for checkpoint signaling after DNA damage. It is ubiquitously expressed, binds to single stranded DNA coated with replication protein A that accumulates at sites of DNA damage and recruits the ataxia telangiectasia and Rad3 related protein (ATR), a checkpoint kinase, to sites of DNA damage and replication stress. Trex1 is required for ATR expression. This gene uses two different open reading frames. The upstream ORF encodes proteins which interact with ATR and localize to intranuclear foci induced by DNA damage and are essential components of the DNA damage checkpoint. The downstream ORF encodes proteins with 3' to 5' exonuclease activity and may be a subunit of human DNA polymerase III. Multiple transcript variants encoding different isoforms have been found. Mutations in this gene result in Aicardi-Goutieres syndrome, chilblain lupus, and Cree encephalitis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 3' repair exonuclease 1; 3'-5' exonuclease TREX1; Deoxyribonuclease III; DNase III; Three-prime repair exonuclease 1
Gene Aliases: AGS1; CRV; DRN3; HERNS; TREX1
UniProt ID: (Human) Q9NSU2
Entrez Gene ID: (Human) 11277
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support