Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive Samples: Mouse eye; Cellular Location: Chromosome, Cytoplasm, Nucleus, cytoskeleton, spindle, telomere
Immunogen sequence: VGAPEEEEEE EEDAGLVAEA EAVAAGWMLD FLCLSLCRAF RDGRSEDFRR TRNSAEAIIH GLSSLTACQL RTIYICQFLT RIAAGKTLDA QFENDERITP LESALMIWGS IEKEHDKLHE EIQNLIKIQA IAVCMENGNF KEAEEVFERI FGDPNSHMPF KSKLLMIISQ KDTFHSFFQH FSYNHMMEKI KSYVNYVLSE KSSTFLMKAA AKVVESKRTR TITSQ
This gene encodes a telomere specific protein which is a component of the telomere nucleoprotein complex. This protein is present at telomeres throughout the cell cycle and functions as an inhibitor of telomerase, acting in cis to limit the elongation of individual chromosome ends. The protein structure contains a C-terminal Myb motif, a dimerization domain near its N-terminus and an acidic N-terminus. Two transcripts of this gene are alternatively spliced products.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ41416; NIMA-interacting protein 2; Telomeric protein Pin2/TRF1; telomeric repeat binding factor (NIMA-interacting) 1; telomeric repeat binding factor 1; Telomeric repeat-binding factor 1; TTAGGG repeat-binding factor 1
Gene Aliases: hTRF1-AS; PIN2; t-TRF1; TERF1; TRBF1; TRF; TRF1
UniProt ID: (Human) P54274
Entrez Gene ID: (Human) 7013, (Rat) 297758
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support