Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Peptide Sequence: MNIHRSTPIT IARYGRSRNK TQDFEELSSI RSAEPSQSFS PNLGSPSPPE TPNLSHCVSC IGKYLLLEPL EGDHVFRAVH LHSGEELVCK VFDISCYQES LAPCFCLSAH SNINQITEII LGETKAYVFF ERSYGDMHSF VRTCKKLREE EAARLFYQIA SAVAHCHDGG LVLRDLKLRK FIFKDEERTR VKLESLEDAY ILRGDDDSLS DKHGCPAYVS PEILNTSGSY SGKAADVWSL GVMLYTMLVG RYPFHDIEPS SLFSKIRRGQ FNIPETLSPK AKCLIRSILR REPSERLTSQ EILDHPWFST DFSVSNSAYG AKEVSDQLVP DVNMEENLDP FFN
This gene encodes one of three members of the Tribbles family. The Tribbles members share a Trb domain, which is homologous to protein serine-threonine kinases, but lacks the active site lysine and probably lacks a catalytic function. The Tribbles proteins interact and modulate the activity of signal transduction pathways in a number of physiological and pathological processes. This Tribbles member induces apoptosis of cells mainly of the hematopoietic origin. It has been identified as a protein up-regulated by inflammatory stimuli in myeloid (THP-1) cells, and also as an oncogene that inactivates the transcription factor C/EBPalpha (CCAAT/enhancer-binding protein alpha) and causes acute myelogenous leukemia. Alternatively spliced transcript variants have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: TRB-2; TRIB2; TRIB2 (HGNC:30809); Tribbles homolog 2
Gene Aliases: C5FW; GS3955; TRB2; TRIB2
UniProt ID: (Human) Q92519
Entrez Gene ID: (Human) 28951
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support