Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: LDAARRDKEA ELQCTQLDLE RKLKLNENAI SRLQANQKSV LVSVSEVKAV AEMQFGELLA AVRKAQANVM LFLEEKEQAA LSQANGIKAH LEYRSAEMEK SKQELERMA
This gene was identified as an estrogen and anti-estrogen regulated gene in epithelial cells stably expressing estrogen receptor. The protein encoded by this gene contains two B box domains and a coiled-coiled region that are characteristic of the B box zinc finger protein family. The proteins of this family have been reported to be involved in a variety of biological processes including cell growth, differentiation and pathogenesis. Expression of this gene was detected in most tissues. Its function, however, has not yet been determined.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase TRIM16; Estrogen-responsive B box protein; tripartite motif-containing 16; Tripartite motif-containing protein 16
Gene Aliases: EBBP; TRIM16
UniProt ID: (Human) O95361
Entrez Gene ID: (Human) 10626
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support