Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: SKPAGADST HKVPVVMLEP IRIKQENSGP PENYDFPVVI VKQESDEESR PQNANYPRSI LTSLLLNSSQ SSTSEETVLR SDAPDSTGDQ PGLHQDNSSN GKSEWLDPSQ KSPLHVGETR KEDDPNEDWC AVCQNGGELL CCEKCPKVFH LSCHVPTLTN FPSGEWICTF CRDLSKPEVE YDCDAPSHNS EKKKTEGLVK LTPIDKRKCE RLLLFLYCHE MSLAFQDPVP LTVPDYYKII KNPMDLSTIK KRLQEDYSMY SKPEDFVADF RLIFQNCAEF NEPDSEVANA GIKLENYFEE LLKNLYPEKR FPKPEFRNES EDNKFSDDSD DDFVQPRKKR LKSIEERQLL K (701-1050 aa encoded by BC028689)
The protein encoded by this gene mediates transcriptional control by interaction with the activation function 2 (AF2) region of several nuclear receptors, including the estrogen, retinoic acid, and vitamin D3 receptors. The protein localizes to nuclear bodies and is thought to associate with chromatin and heterochromatin-associated factors. The protein is a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains - a RING, a B-box type 1 and a B-box type 2 - and a coiled-coil region. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ab2-427; E3 ubiquitin-protein ligase TRIM24; RING finger protein 82; RING-type E3 ubiquitin transferase TIF1-alpha; TIF1-alpha; Transcription intermediary factor 1-alpha; transcriptional intermediary factor 1; transcriptional intermediary factor 1, alpha; tripartite motif-containing 24; Tripartite motif-containing protein 24
Gene Aliases: A130082H20Rik; AI447469; D430004I05Rik; hTIF1; PTC6; RNF82; TF1A; TIF1; TIF1-alpha; TIF1A; TIF1ALPHA; TRIM24
UniProt ID: (Human) O15164, (Mouse) Q64127
Entrez Gene ID: (Human) 8805, (Rat) 500084, (Mouse) 21848
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support