Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MAELCPLAEE LSCSICLEPF KEPVTTPCGH NFCGSCLNET WAVQGSPYLC PQCRAVYQAR PQLHKNTVLC NVVEQFLQAD LAREPPADVW TPPARASAPS PNAQVACDHC LKEAAVKTCL VCMASFCQEH LQPHFDSPAF QDHPLQPPVR DLLRRKCSQH NRLREFFCPE HSECICHICL VEHKTCSPAS LSQASADLEA TLRHKLTVMY SQINGASRAL DDVRNRQQDV RMTANRKVEQ LQQEYTEMKA LLDASETTST RKIKEEEKRV NSKFDTIYQI LLKKKSEIQT LKEEIEQSLT KRDEFEFLEK ASKLRGISTK PVYIPEVELN HKLIKGIHQS TIDLKNELKQ CIGRLQELTP SSGDPGEHDP ASTHKSTRPV KKVSKEEKKS KKPPPVPALP SKLPTFGAPE QLVDLKQAGL EAAAKATSSH PNSTSLKAKV LETFLAKSRP ELLEYYIKVI LDYNTAHNKV ALSECYTVAS VAEMPQNYRP HPQRFTYCSQ VLGLHCYKKG IHYWEVELQK NNFCGVGICY GSMNRQGPES RLGRNSASWC VEWFNTKISA WHNNVEKTLP STKATRVGVL LNCDHGFVIF FAVADKVHLM YKFRVDFTEA LYPAFWVFSA GATLSICSPK
TRIM25, also known as estrogen responsive finger protein (EFP) is a member of the RING finger-B box-coiled coil family and is a downstream target of estrogen receptor a. Mice lacking this gene displayed an underdeveloped uterus and reduced estrogen responsiveness demonstrating that this protein is essential for estrogen dependent proliferation. TRIM25 expression is increased in breast carcinoma where it promotes the growth of breast tumor by functioning as an E3 ubiquitin ligase that targets the negative cell cycle checkpoint 14-3-3s. Recent reports have indicated that TRIM25 E3 ubiquitin ligase is essential for the retinoic-acid-inducible gene 1 (RIG-1) signaling pathway to elicit host antiviral innate immunity.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin/ISG15 ligase TRIM25; Estrogen-responsive finger protein; RING finger protein 147; RING-type E3 ubiquitin transferase; RING-type E3 ubiquitin transferase TRIM25; tripartite motif protein TRIM25; tripartite motif-containing 25; Tripartite motif-containing protein 25; Ubiquitin/ISG15-conjugating enzyme TRIM25; Zinc finger protein 147; zinc finger protein 147 (estrogen-responsive finger protein); zinc finger protein-147
Gene Aliases: EFP; RNF147; TRIM25; Z147; ZNF147
UniProt ID: (Human) Q14258
Entrez Gene ID: (Human) 7706
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support