Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TRIM31 encodes for a member of the tripartite motif (TRIM) family. The TRIM motif includes three zinc-binding domains, a RING, a B-box type 1 and a B-box type 2, and a coiled-coil region. The protein localizes to both the cytoplasm and the nucleus.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: E3 ubiquitin-protein ligase TRIM31; tripartite motif-containing 31; Tripartite motif-containing protein 31
Gene Aliases: C6orf13; HCG1; HCGI; RNF; TRIM31
UniProt ID: (Human) Q5SS28
Entrez Gene ID: (Human) 11074
Molecular Function:
ubiquitin-protein ligase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support